4-Fluor-3-(methylcarbamoyl)phenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Fluoro-3-(methylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid

PC5016-250MG PC5016-1G PC5016-5G
APOSPC5016-250MGEA 43.5 EUR
APOSPC5016-250MG APOSPC5016-1G APOSPC5016-5G
4-Fluor-3-(methylcarbamoyl)phenylboronsäure 96%
4-Fluor-3-(methylcarbamoyl)phenylboronsäure
Formel: C₈H₉BFNO₃
Molecular Weight: 196.97 g/mol
Schmelzpunkt: 226…228 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235038
CAS-Nummer: 874219-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR