(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2-Difluoro-1,3-benzodioxole-5-boronic acid, (2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid

PC501601-1G
APOSPC501601-1GEA 91.5 EUR
APOSPC501601-1G
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid
Formel: C₇H₅BF₂O₄
Molecular Weight: 201.92 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08706236
CAS-Nummer: 190903-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR