2-Fluoro-4-methyl-5-nitroaniline 97%

Lieferant: Apollo Scientific

PC501611-1G
APOSPC501611-1GEA 0 EUR
APOSPC501611-1G
2-Fluoro-4-methyl-5-nitroaniline 97%
2-Fluoro-4-methyl-5-nitroaniline
Formel: C₇H₇FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD15527685
CAS-Nummer: 259860-00-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR