4-Fluoro-2-nitrobenzenesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC501630-1G
APOSPC501630-1GEA
157
EUR
APOSPC501630-1G
4-Fluoro-2-nitrobenzenesulphonyl chloride
4-Fluoro-2-nitrobenzenesulphonyl chloride
Formel:
C₆H₃ClFNO₄S Siedepunkt: 150 - 155 °C Schmelzpunkt: 44 - 47 °C |
MDL:
MFCD03618461 CAS-Nummer: 568586-10-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...