Methyl 5-fluoro-2-formylbenzoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501659-250MG
APOSPC501659-250MGEA
195
EUR
APOSPC501659-250MG
Methyl 5-fluoro-2-formylbenzoate 95%
Methyl 5-fluoro-2-formylbenzoate
Formel:
C₉H₇FO₃ Molecular Weight: 182,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226921 CAS-Nummer: 1194374-71-4 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...