4-(Pentafluorothio)benzeneboronic acid, pinacol ester 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-phenylsulphur pentafluoride
PC501675-1G
PC501675-5G
APOSPC501675-1GEA
575
EUR
APOSPC501675-1G
APOSPC501675-5G
4-(Pentafluorothio)benzeneboronic acid, pinacol ester 95%
4-(Pentafluorothio)benzeneboronic acid, pinacol ester
Formel:
C₁₂H₁₆BF₅O₂S Molecular Weight: 330.122 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20489194 CAS-Nummer: 1286278-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...