4-(Pentafluorothio)benzeneboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-phenylsulphur pentafluoride

PC501675-1G PC501675-5G
APOSPC501675-1GEA 575 EUR
APOSPC501675-1G APOSPC501675-5G
4-(Pentafluorothio)benzeneboronic acid, pinacol ester 95%
4-(Pentafluorothio)benzeneboronic acid, pinacol ester
Formel: C₁₂H₁₆BF₅O₂S
Molecular Weight: 330.122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20489194
CAS-Nummer: 1286278-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR