3-(Butylcarbamoyl)-4-fluorobenzeneboronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
3-(N-Butylcarbamoyl)-4-fluorophenylboronic acid
PC5017-1G
APOSPC5017-1GEA
280
EUR
APOSPC5017-1G
3-(Butylcarbamoyl)-4-fluorobenzeneboronic acid 98%
3-(Butylcarbamoyl)-4-fluorobenzeneboronic acid
Formel:
C₁₁H₁₅BFNO₃ Molecular Weight: 239,051 g/mol Schmelzpunkt: 242…244 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235042 CAS-Nummer: 874219-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...