4-(Difluoromethoxy)-2-nitroaniline 97%

Lieferant: Apollo Scientific

PC501715-250MG
APOSPC501715-250MGEA 36.4 EUR
APOSPC501715-250MG
4-(Difluoromethoxy)-2-nitroaniline 97%
4-(Difluoromethoxy)-2-nitroaniline
Formel: C₇H₆F₂N₂O₃
Molecular Weight: 204,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037370
CAS-Nummer: 97963-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR