2-Amino-5-fluoro-4-nitrophenol 97%

Lieferant: Apollo Scientific

PC501727-250MG
APOSPC501727-250MGEA 244 EUR
APOSPC501727-250MG
2-Amino-5-fluoro-4-nitrophenol 97%
2-Amino-5-fluoro-4-nitrophenol
Formel: C₆H₅FN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28967930
CAS-Nummer: 1850312-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR