3-Bromo-2-(difluoromethoxy)aniline 95%

Lieferant: Apollo Scientific
Gefahr

PC501742-1G PC501742-250MG
APOSPC501742-1GEA 0 EUR
APOSPC501742-1G APOSPC501742-250MG
3-Bromo-2-(difluoromethoxy)aniline 95%
3-Bromo-2-(difluoromethoxy)aniline
Formel: C₇H₆BrF₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18390859
CAS-Nummer: 259243-50-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR