5-Bromo-2,3,4-trifluorobenzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-2,3,4-trifluorophenyl)boronic acid, 5-Bromo-2,3,4-trifluorophenylboronic acid

PC50182-1G PC50182-250MG PC50182-5G
APOSPC50182-1GEA 522 EUR
APOSPC50182-1G APOSPC50182-250MG APOSPC50182-5G
5-Bromo-2,3,4-trifluorobenzeneboronic acid 95%
5-Bromo-2,3,4-trifluorobenzeneboronic acid
Formel: C₆H₃BBrF₃O₂
Molecular Weight: 254.797 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12032026
CAS-Nummer: 2377611-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR