5-Bromo-3-iodo-2-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific

PC50184-1G PC50184-250MG PC50184-5G
APOSPC50184-1GEA 0 EUR
APOSPC50184-1G APOSPC50184-250MG APOSPC50184-5G
5-Bromo-3-iodo-2-(trifluoromethyl)pyridine 95%
5-Bromo-3-iodo-2-(trifluoromethyl)pyridine
Formel: C₆H₂BrF₃IN
Lagertemperatur: Raumtemperatur
MDL: MFCD23382317
CAS-Nummer: 1408279-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR