3-Bromo-2,5-difluorobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC501845-1G PC501845-250MG PC501845-5G
APOSPC501845-1GEA 219 EUR
APOSPC501845-1G APOSPC501845-250MG APOSPC501845-5G
3-Bromo-2,5-difluorobenzoic acid 95%
3-Bromo-2,5-difluorobenzoic acid
Formel: C₇H₃BrF₂O₂
Molecular Weight: 236.998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24107184
CAS-Nummer: 1520538-81-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR