3-Bromo-2,5-difluorobenzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501845-1G
PC501845-250MG
PC501845-5G
APOSPC501845-1GEA
219
EUR
APOSPC501845-1G
APOSPC501845-250MG
APOSPC501845-5G
3-Bromo-2,5-difluorobenzoic acid 95%
3-Bromo-2,5-difluorobenzoic acid
Formel:
C₇H₃BrF₂O₂ Molecular Weight: 236.998 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24107184 CAS-Nummer: 1520538-81-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...