3-Nitro-4-(trifluoromethoxy)benzonitrile

Lieferant: Apollo Scientific

PC501852-1G
APOSPC501852-1GEA 0 EUR
APOSPC501852-1G
3-Nitro-4-(trifluoromethoxy)benzonitrile
3-Nitro-4-(trifluoromethoxy)benzonitrile
Formel: C₈H₃F₃N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09745165
CAS-Nummer: 1214360-38-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR