2-Bromo-3,5-difluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC501854-1G PC501854-5G PC501854-25G
APOSPC501854-1GEA 21.9 EUR
APOSPC501854-1G APOSPC501854-5G APOSPC501854-25G
2-Bromo-3,5-difluorobenzoic acid
2-Bromo-3,5-difluorobenzoic acid
Formel: C₇H₃BrF₂O₂
Molecular Weight: 236.998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835211
CAS-Nummer: 651027-01-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR