5-Bromo-2-(trifluoromethyl)pyridine-3-carbonitrile 95%

Lieferant: Apollo Scientific

PC50189-1G PC50189-250MG
APOSPC50189-1GEA 0 EUR
APOSPC50189-1G APOSPC50189-250MG
5-Bromo-2-(trifluoromethyl)pyridine-3-carbonitrile 95%
5-Bromo-2-(trifluoromethyl)pyridine-3-carbonitrile
Formel: C₇H₂BrF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11848373
CAS-Nummer: 1245914-98-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR