4-(Difluoromethoxy)-3-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC501893-1G PC501893-250MG PC501893-5G
APOSPC501893-1GEA 24.8 EUR
APOSPC501893-1G APOSPC501893-250MG APOSPC501893-5G
4-(Difluoromethoxy)-3-fluoroaniline
4-(Difluoromethoxy)-3-fluoroaniline
Formel: C₇H₆F₃NO
Lagertemperatur: Kühlschrank
MDL: MFCD11621754
CAS-Nummer: 83190-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR