3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (3-(Ethylcarbamoyl)-4-fluorophenyl)boronic acid, 3-(Ethylcarbamoyl)-4-fluorophenylboronic acid

PC5019-1G
APOSPC5019-1GEA 306 EUR
APOSPC5019-1G
3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid 98%
3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid
Formel: C₉H₁₁BFNO₃
Molecular Weight: 210,998 g/mol
Schmelzpunkt: 260…262 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235039
CAS-Nummer: 874219-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR