2-(Difluoromethoxy)-6-fluoroaniline
Lieferant: Apollo Scientific
|
Achtung
|
PC501904-1G
PC501904-250MG
PC501904-5G
APOSPC501904-1GEA
412
EUR
APOSPC501904-1G
APOSPC501904-250MG
APOSPC501904-5G
2-(Difluoromethoxy)-6-fluoroaniline
2-(Difluoromethoxy)-6-fluoroaniline
Formel:
C₇H₆F₃NO Lagertemperatur: Kühlschrank |
MDL:
MFCD18385074 CAS-Nummer: 1261839-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...