3-Bromo-2,5-difluorobenzonitrile 98%

Lieferant: Apollo Scientific
Gefahr

PC501939-250MG
APOSPC501939-250MGEA 162 EUR
APOSPC501939-250MG
3-Bromo-2,5-difluorobenzonitrile 98%
3-Bromo-2,5-difluorobenzonitrile
Formel: C₇H₂BrF₂N
Molecular Weight: 217,998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27923485
CAS-Nummer: 1638487-41-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR