5-(Difluoromethoxy)-2-fluoronitrobenzene 95%

Lieferant: Apollo Scientific

PC501967-1G PC501967-250MG PC501967-5G
APOSPC501967-1GEA 575 EUR
APOSPC501967-1G APOSPC501967-250MG APOSPC501967-5G
5-(Difluoromethoxy)-2-fluoronitrobenzene 95%
5-(Difluoromethoxy)-2-fluoronitrobenzene
Formel: C₇H₄F₃NO₃
Molecular Weight: 207.107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698454
CAS-Nummer: 57213-30-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR