3-Bromo-2,6-difluorobenzyl bromide

Lieferant: Apollo Scientific

PC501973-1G
APOSPC501973-1GEA 574 EUR
APOSPC501973-1G
3-Bromo-2,6-difluorobenzyl bromide
3-Bromo-2,6-difluorobenzyl bromide
Formel: C₇H₄Br₂F₂
Molecular Weight: 285,911 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27950792
CAS-Nummer: 438050-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR