2-(Difluoromethoxy)-4-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC501980-1G PC501980-5G
APOSPC501980-1GEA 241 EUR
APOSPC501980-1G APOSPC501980-5G
2-(Difluoromethoxy)-4-fluoroaniline
2-(Difluoromethoxy)-4-fluoroaniline
Formel: C₇H₆F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD04969924
CAS-Nummer: 832740-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR