2-Bromo-3,4-difluorobenzaldehyde

Lieferant: Apollo Scientific
Achtung

PC501996-1G PC501996-250MG
APOSPC501996-1GEA 102 EUR
APOSPC501996-1G APOSPC501996-250MG
2-Bromo-3,4-difluorobenzaldehyde
2-Bromo-3,4-difluorobenzaldehyde
Formel: C₇H₃BrF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD28669844
CAS-Nummer: 1578156-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR