2-(Difluoromethoxy)-3-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC501999-1G PC501999-250MG
APOSPC501999-1GEA 0 EUR
APOSPC501999-1G APOSPC501999-250MG
2-(Difluoromethoxy)-3-fluoroaniline
2-(Difluoromethoxy)-3-fluoroaniline
Formel: C₇H₆F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD14698441
CAS-Nummer: 153468-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR