3-Iodo-5-(trifluoromethyl)phenacyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
PC502024-1G
PC502024-250MG
PC502024-5G
APOSPC502024-1GEA
0
EUR
APOSPC502024-1G
APOSPC502024-250MG
APOSPC502024-5G
3-Iodo-5-(trifluoromethyl)phenacyl bromide
3-Iodo-5-(trifluoromethyl)phenacyl bromide
Formel:
C₉H₅BrF₃IO Lagertemperatur: Raumtemperatur |
MDL:
MFCD28060517 CAS-Nummer: 1823897-02-4 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...