2-(Chloromethyl)-5-fluoro-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

PC502040-250MG
APOSPC502040-250MGEA 140 EUR
APOSPC502040-250MG
2-(Chloromethyl)-5-fluoro-1,3-benzoxazole
2-(Chloromethyl)-5-fluoro-1,3-benzoxazole
Formel: C₈H₅ClFNO
Molecular Weight: 185,583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696613
CAS-Nummer: 143708-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR