3-Bromo-4,5-difluorobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC502061-1G
APOSPC502061-1GEA 316 EUR
APOSPC502061-1G
3-Bromo-4,5-difluorobenzyl alcohol
3-Bromo-4,5-difluorobenzyl alcohol
Formel: C₇H₅BrF₂O
Molecular Weight: 223.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28737916
CAS-Nummer: 1143502-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR