Methyl-2-bromo-3-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific

PC502066-1G PC502066-250MG PC502066-500MG PC502066-2.5G
APOSPC502066-1GEA 0 EUR
APOSPC502066-1G APOSPC502066-250MG APOSPC502066-500MG APOSPC502066-2.5G
Methyl-2-bromo-3-(trifluoromethyl)benzoate
Methyl-2-bromo-3-(trifluoromethyl)benzoate
Formel: C₉H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14697994
CAS-Nummer: 1214362-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR