Methyl 2,6-difluoro-3-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC502071-1G PC502071-250MG PC502071-5G
APOSPC502071-1GEA 335 EUR
APOSPC502071-1G APOSPC502071-250MG APOSPC502071-5G
Methyl 2,6-difluoro-3-(trifluoromethyl)benzoate
Methyl 2,6-difluoro-3-(trifluoromethyl)benzoate
Formel: C₉H₅F₅O₂
Lagertemperatur: Kühlschrank
MDL: MFCD30481112
CAS-Nummer: 2089258-73-9
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR