2-(Trifluoromethyl)pyrimidine-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC50209-25G
APOSPC50209-25GEA 521 EUR
APOSPC50209-25G
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
Formel: C₆H₃F₃N₂O₂
Schmelzpunkt: 170…175 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09054851
CAS-Nummer: 306960-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR