3-Iodo-4-(trifluoromethyl)benzonitrile

Lieferant: Apollo Scientific
Gefahr

PC502103-1G PC502103-250MG
APOSPC502103-1GEA 364 EUR
APOSPC502103-1G APOSPC502103-250MG
3-Iodo-4-(trifluoromethyl)benzonitrile
3-Iodo-4-(trifluoromethyl)benzonitrile
Formel: C₈H₃F₃IN
Lagertemperatur: Kühlschrank
MDL: MFCD18398161
CAS-Nummer: 1261482-46-5
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR