2,4-Difluoro-3-(trifluoromethoxy)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC502108-1G PC502108-250MG
APOSPC502108-1GEA 290 EUR
APOSPC502108-1G APOSPC502108-250MG
2,4-Difluoro-3-(trifluoromethoxy)benzoic acid
2,4-Difluoro-3-(trifluoromethoxy)benzoic acid
Formel: C₈H₃F₅O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07787527
CAS-Nummer: 1119454-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR