2,4-Dichloro-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC50211-1G
APOSPC50211-1GEA 357 EUR
APOSPC50211-1G
2,4-Dichloro-6-(trifluoromethyl)pyridine
2,4-Dichloro-6-(trifluoromethyl)pyridine
Formel: C₆H₂Cl₂F₃N
Molecular Weight: 215,988 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698621
CAS-Nummer: 39891-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR