4-chloro-6-fluoro-2-(trifluoromethyl)quinazoline 95+%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-6-fluoro-2-(trifluoromethyl)quinazoline

PC502110-250MG
APOSPC502110-250MGEA 81.2 EUR
APOSPC502110-250MG
4-chloro-6-fluoro-2-(trifluoromethyl)quinazoline 95+%
4-chloro-6-fluoro-2-(trifluoromethyl)quinazoline
Formel: C₉H₃ClF₄N₂
Molecular Weight: 250,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954902
CAS-Nummer: 959238-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR