Methyl 2-bromo-3-fluorobenzoate

Lieferant: Apollo Scientific
Achtung

PC502130-1G PC502130-250MG PC502130-5G
APOSPC502130-1GEA 95.7 EUR
APOSPC502130-1G APOSPC502130-250MG APOSPC502130-5G
Methyl 2-bromo-3-fluorobenzoate
Methyl 2-bromo-3-fluorobenzoate
Formel: C₈H₆BrFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10000921
CAS-Nummer: 647020-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR