Methyl 3-iodo-5-(trifluoromethyl)benzoate 95%

Lieferant: Apollo Scientific
Achtung

PC502131-1G PC502131-250MG PC502131-5G
APOSPC502131-1GEA 0 EUR
APOSPC502131-1G APOSPC502131-250MG APOSPC502131-5G
Methyl 3-iodo-5-(trifluoromethyl)benzoate 95%
Methyl 3-iodo-5-(trifluoromethyl)benzoate
Formel: C₉H₆F₃IO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11976042
CAS-Nummer: 161622-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR