4-Bromo-2-chloro-3-fluoroaniline 99%

Lieferant: Apollo Scientific
Achtung

PC502134-1G PC502134-250MG PC502134-5G
APOSPC502134-1GEA 174 EUR
APOSPC502134-1G APOSPC502134-250MG APOSPC502134-5G
4-Bromo-2-chloro-3-fluoroaniline 99%
4-Bromo-2-chloro-3-fluoroaniline
Formel: C₆H₄BrClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD19381659
CAS-Nummer: 1349719-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR