3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid 97%

Lieferant: Apollo Scientific

Synonyme: (3-Fluoro-2-(hydroxymethyl)phenyl)boronic acid, 3-Fluoro-2-(hydroxymethyl)phenylboronic acid

PC502136-1G PC502136-5G
APOSPC502136-1GEA 87 EUR
APOSPC502136-1G APOSPC502136-5G
3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid 97%
3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid
Formel: C₇H₈BFO₃
Molecular Weight: 169.946 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13181611
CAS-Nummer: 1246633-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR