3-Iodo-5-(trifluoromethyl)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
PC502146-1G
PC502146-250MG
PC502146-5G
APOSPC502146-1GEA
1400
EUR
APOSPC502146-1G
APOSPC502146-250MG
APOSPC502146-5G
3-Iodo-5-(trifluoromethyl)benzyl bromide
3-Iodo-5-(trifluoromethyl)benzyl bromide
Formel:
C₈H₅BrF₃I Lagertemperatur: Raumtemperatur |
MDL:
MFCD18398193 CAS-Nummer: 1261795-42-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...