5-Chloro-4-fluoro-2-methylaniline 97%
Lieferant: Apollo Scientific
PC502149-250MG
APOSPC502149-250MGEA
36.4
EUR
APOSPC502149-250MG
5-Chloro-4-fluoro-2-methylaniline 97%
5-Chloro-4-fluoro-2-methylaniline
Formel:
C₇H₇ClFN Molecular Weight: 159,589 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16817674 CAS-Nummer: 1242339-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...