2-(Difluoromethoxy)-6-fluorobenzaldehyde 95%

Lieferant: Apollo Scientific

PC502196-1G
APOSPC502196-1GEA 93.1 EUR
APOSPC502196-1G
2-(Difluoromethoxy)-6-fluorobenzaldehyde 95%
2-(Difluoromethoxy)-6-fluorobenzaldehyde
Formel: C₈H₅F₃O₂
Molecular Weight: 190,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698497
CAS-Nummer: 1214333-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR