3-Benzylcarbamoyl-4-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Benzylcarbamoyl)-4-fluorophenylboronic acid

PC5022-1G
APOSPC5022-1GEA 205 EUR
APOSPC5022-1G
3-Benzylcarbamoyl-4-fluorphenylboronsäure 98%
3-Benzylcarbamoyl-4-fluorphenylboronsäure
Formel: C₁₄H₁₃BFNO₃
Schmelzpunkt: 237…239 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235041
CAS-Nummer: 874219-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR