5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
PC50223-100MG
PC50223-1G
PC50223-250MG
APOSPC50223-100MGEA
134
EUR
APOSPC50223-100MG
APOSPC50223-1G
APOSPC50223-250MG
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
Formel:
C₁₀H₁₀F₃N·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD08544284 CAS-Nummer: 215788-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...