5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline

Lieferant: Apollo Scientific
Achtung

PC50223-100MG PC50223-1G PC50223-250MG
APOSPC50223-100MGEA 134 EUR
APOSPC50223-100MG APOSPC50223-1G APOSPC50223-250MG
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
Formel: C₁₀H₁₀F₃N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08544284
CAS-Nummer: 215788-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR