4-Bromo-7-fluorobenzo[b]thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
Achtung
|
PC502239-1G
APOSPC502239-1GEA
899
EUR
APOSPC502239-1G
4-Bromo-7-fluorobenzo[b]thiophene-2-carboxylic acid
4-Bromo-7-fluorobenzo[b]thiophene-2-carboxylic acid
Formel:
C₉H₄BrFO₂S Molecular Weight: 275,09 g/mol |
MDL:
MFCD13180864 CAS-Nummer: 360576-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...