4-Bromo-6-fluorobenzo[b]thiophene-2-carboxylic acid ≥98%

Lieferant: Apollo Scientific

PC502240-100MG
APOSPC502240-100MGEA 144 EUR
APOSPC502240-100MG
4-Bromo-6-fluorobenzo[b]thiophene-2-carboxylic acid ≥98%
4-Bromo-6-fluorobenzo[b]thiophene-2-carboxylic acid
Formel: C₉H₄BrFO₂S
Molecular Weight: 275,094 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13180839
CAS-Nummer: 826995-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR