2-Ethynyl-4-fluorobenzaldehyde 97%

Lieferant: Apollo Scientific

PC502251-250MG
APOSPC502251-250MGEA 40.6 EUR
APOSPC502251-250MG
2-Ethynyl-4-fluorobenzaldehyde 97%
2-Ethynyl-4-fluorobenzaldehyde
Formel: C₉H₅FO
Lagertemperatur: Raumtemperatur
MDL: MFCD18824363
CAS-Nummer: 749874-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR